Quizartinib ≥98%

Supplier: Ambeed
Warning

Synonyms: AC220, 1-(5-(tert-Butyl)isoxazol-3-yl)-3-(4-(7-(2-morpholinoethoxy)benzo[d]imidazo[2,1-b]thiazol-2-yl)phenyl)urea

A475775-100MG A475775-10MG A475775-250MG A475775-50MG
77602-066EA 82 USD
77602-066 77613-486 77613-488 77613-490
Quizartinib ≥98%
Quizartinib

Precaución: for Research Use Only

Formula: C₂₉H₃₂N₆O₄S
MW: 560.7 g/mol
MDL Number: MFCD18074524
CAS Number: 950769-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR