Methyl-3-methylbenzo[b]thiophene-2-carboxylate ≥98%

Supplier: Ambeed
Warning

A481687-1G A481687-5G
77686-272EA 30 USD
77686-272 77901-628
Methyl-3-methylbenzo[b]thiophene-2-carboxylate ≥98%
Methyl 3-methylbenzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₀O₂S
MDL Number: MFCD02683110
CAS Number: 3133-81-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR