N-(Pyridin-3-yl)-4-(3-((5-(trifluoromethyl)pyridin-2-yl)oxy)benzyl)piperidine-1-carboxamide 99+%

Supplier: Ambeed
Danger

Synonyms: PF-3845

A475760-10MG A475760-1MG A475760-5MG
77613-480EA 40.17 USD
77613-480 77613-482 77613-484
N-(Pyridin-3-yl)-4-(3-((5-(trifluoromethyl)pyridin-2-yl)oxy)benzyl)piperidine-1-carboxamide 99+%
N-(Pyridin-3-yl)-4-(3-((5-(trifluoromethyl)pyridin-2-yl)oxy)benzyl)piperidine-1-carboxamide

Precaución: for Research Use Only

Formula: C₂₄H₂₃F₃N₄O₂
MW: 456.46 g/mol
MDL Number: MFCD18382105
CAS Number: 1196109-52-0
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR