Boc-D-Nle-OH ≥98%

Supplier: Ambeed
Warning

Synonyms: (R)-2-((tert-Butoxycarbonyl)amino)hexanoic acid

A475746-1G A475746-25G A475746-5G
77613-474EA 40.17 USD
77613-474 77613-476 77613-478
Boc-D-Nle-OH ≥98%
Boc-D-Nle-OH

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₄
MW: 231.292 g/mol
MDL Number: MFCD00063168
CAS Number: 55674-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR