(2-Cyanopyridin-4-yl)boronic acid 95+%

Supplier: Ambeed
Warning

A475639-100MG A475639-250MG A475639-5G
77884-211EA 54 USD
77884-211 77884-212 77884-213
(2-Cyanopyridin-4-yl)boronic acid 95+%
(2-Cyanopyridin-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₅BN₂O₂
MDL Number: MFCD10696640
CAS Number: 903513-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR