Isoquercitrin ≥95%

Supplier: Ambeed
Warning

Synonyms: 3-(((2S,3R,4R,5R)-5-((R)-1,2-Dihydroxyethyl)-3,4-dihydroxytetrahydrofuran-2-yl)oxy)-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-4H-chromen-4-one, Quercetin-3-Glucoside, Trifoliin, 3,3',4',5,7-Pentahydroxyflavone-3-β-Glucoside

A475548-10MG A475548-1MG A475548-5MG
77613-468EA 39.14 USD
77613-468 77613-470 77613-472
Isoquercitrin ≥95%
Isoquercitrin

Precaución: for Research Use Only

Formula: C₂₁H₂₀O₁₂
MW: 464.38 g/mol
MDL Number: MFCD29050128
CAS Number: 21637-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR