4-(4-Chlorophenyl)-1-(4-(4-fluorophenyl)-4-oxobutyl)piperidin-4-yl decanoate ≥97%

Supplier: Ambeed
Warning

Synonyms: Haloperidol decanoate

A481461-1MG A481461-5MG A481461-10MG
77617-082EA 6.18 USD
77617-082 77617-084 77748-995
4-(4-Chlorophenyl)-1-(4-(4-fluorophenyl)-4-oxobutyl)piperidin-4-yl decanoate ≥97%
4-(4-Chlorophenyl)-1-(4-(4-fluorophenyl)-4-oxobutyl)piperidin-4-yl decanoate

Precaución: for Research Use Only

Formula: C₃₁H₄₁ClFNO₃
MW: 530.11 g/mol
MDL Number: MFCD00866706
CAS Number: 74050-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR