Methyl-2,3-dihydro-1,4-benzodioxine-5-carboxylate ≥95%

Supplier: Ambeed
Warning

A475341-5G A475341-1G A475341-250MG A475341-100MG
77673-228EA 906 USD
77673-228 77884-209 77884-210 77884-208
Methyl-2,3-dihydro-1,4-benzodioxine-5-carboxylate ≥95%
Methyl-2,3-dihydro-1,4-benzodioxine-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MDL Number: MFCD01546256
CAS Number: 214894-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR