1,4-Dibromo-2,5-diethylbenzene ≥98%

Supplier: Ambeed
Warning

A481165-5G A481165-1G
77622-336EA 100.94 USD
77622-336 77867-822
1,4-Dibromo-2,5-diethylbenzene ≥98%
1,4-Dibromo-2,5-diethylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂Br₂
MW: 292.0103 g/mol
MDL Number: MFCD04038416
CAS Number: 40787-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR