(4-Fluoro-3-methylphenyl)acetic acid ≥98%

Supplier: Ambeed
Warning

A481135-250MG A481135-1G
77673-246EA 45 USD
77673-246 77872-794
(4-Fluoro-3-methylphenyl)acetic acid ≥98%
(4-Fluoro-3-methylphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₂
MDL Number: MFCD09832241
CAS Number: 1000520-92-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR