(3R,4S)-tert-Butyl 3-amino-4-fluoropyrrolidine-1-carboxylate ≥95%

Supplier: Ambeed
Warning

A474973-100MG A474973-250MG
77886-235EA 71 USD
77886-235 77886-236
(3R,4S)-tert-Butyl 3-amino-4-fluoropyrrolidine-1-carboxylate ≥95%
(3R,4S)-tert-Butyl 3-amino-4-fluoropyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₇FN₂O₂
MDL Number: MFCD18071735
CAS Number: 1009075-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR