N-Ethylpyrimidin-2-amine ≥95%

Supplier: Ambeed
Warning

A474865-1G A474865-250MG
77686-257EA 332 USD
77686-257 77686-258
N-Ethylpyrimidin-2-amine ≥95%
N-Ethylpyrimidin-2-amine

Precaución: for Research Use Only

Formula: C₆H₉N₃
MW: 123.15 g/mol
MDL Number: MFCD11120076
CAS Number: 66131-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR