4-tert-Butyl-2-hydroxybenzaldehyde ≥95%

Supplier: Ambeed
Warning

Synonyms: 4-(tert-Butyl)-2-hydroxybenzaldehyde

A480531-1G A480531-5G
77572-510EA 37.08 USD
77572-510 77602-074
4-tert-Butyl-2-hydroxybenzaldehyde ≥95%
4-tert-Butyl-2-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.228 g/mol
MDL Number: MFCD06412558
CAS Number: 66232-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR