1-(2-Bromo-3-chlorophenyl)ethanone ≥98%

Supplier: Ambeed
Warning

A474329-1G A474329-5G
77686-255EA 31 USD
77686-255 77686-256
1-(2-Bromo-3-chlorophenyl)ethanone ≥98%
1-(2-Bromo-3-chlorophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₆BrClO
MW: 233.49 g/mol
MDL Number: MFCD18390297
CAS Number: 161957-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR