Dimethyl 2-(2-Pyrimidyl)malonate ≥98%

Supplier: Ambeed
Warning

A473966-250MG A473966-1G
77882-200EA 6 USD
77882-200 77882-199
Dimethyl 2-(2-Pyrimidyl)malonate ≥98%
Dimethyl 2-(2-Pyrimidyl)malonate

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O₄
MDL Number: MFCD12198886
CAS Number: 93271-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR