(R)-tert-Butyl (1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethyl)carbamate ≥95%

Supplier: Ambeed
Warning

A470592-250MG A470592-100MG
77748-970EA 195 USD
77748-970 77748-969
(R)-tert-Butyl (1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethyl)carbamate ≥95%
(R)-tert-Butyl (1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₉H₃₀BNO₄
MDL Number: MFCD18632958
CAS Number: 578729-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR