Methyl 3-bromo-4,5-diaminobenzoate ≥98%

Supplier: Ambeed
Warning

A470533-1G A470533-250MG A470533-5G
77892-950EA 21 USD
77892-950 77892-951 77892-952
Methyl 3-bromo-4,5-diaminobenzoate ≥98%
Methyl 3-bromo-4,5-diaminobenzoate

Precaución: for Research Use Only

Formula: C₈H₉BrN₂O₂
MDL Number: MFCD18072579
CAS Number: 1245643-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR