(R)-1-(3,5-Dichloropyridin-4-yl)ethanol ≥98%

Supplier: Ambeed
Warning

A470505-100MG A470505-250MG
77901-306EA 41 USD
77901-306 77901-338
(R)-1-(3,5-Dichloropyridin-4-yl)ethanol ≥98%
(R)-1-(3,5-Dichloropyridin-4-yl)ethanol

Precaución: for Research Use Only

Formula: C₇H₇Cl₂NO
MDL Number: MFCD22665798
CAS Number: 1254473-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR