3-Imidazol-1-ylphenylamine ≥98%

Supplier: Ambeed
Warning

A470441-1G A470441-5G
77890-952EA 47 USD
77890-952 77890-953
3-Imidazol-1-ylphenylamine ≥98%
3-Imidazol-1-ylphenylamine

Precaución: for Research Use Only

Formula: C₉H₉N₃
MDL Number: MFCD09258808
CAS Number: 112677-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR