3-Bromo-2,5-difluoroaniline ≥95%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
A470284-1G
A470284-100MG
A470284-250MG
77684-267EA
89
USD
77684-267
77890-950
77890-951
3-Bromo-2,5-difluoroaniline ≥95%, Ambeed.Inc
3-Bromo-2,5-difluoroaniline
Precaución: For research use only.
Formula:
C₆H₄BrF₂N |
MDL Number:
MFCD22573901 CAS Number: 1269232-99-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...