N6,N6-Dimethyladenosine ≥97%

Supplier: Ambeed
Warning

A469786-1G A469786-250MG
77669-108EA 163 USD
77669-108 77669-110
N6,N6-Dimethyladenosine ≥97%
N6,N6-Dimethyladenosine

Precaución: for Research Use Only

Formula: C₁₂H₁₇N₅O₄
MDL Number: MFCD00022822
CAS Number: 2620-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR