2-(2-Carboxyethyl)-4-methyl-5-propylfuran-3-carboxylic acid ≥97%

Supplier: Ambeed
Warning

Synonyms: CMPF

A472725-1MG A472725-10MG
77617-056EA 61.8 USD
77617-056 77748-982
2-(2-Carboxyethyl)-4-methyl-5-propylfuran-3-carboxylic acid ≥97%
2-(2-Carboxyethyl)-4-methyl-5-propylfuran-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₅
MW: 240.25 g/mol
CAS Number: 86879-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR