RuCl[(R,R)-Fsdpen](p-cymene) ≥98%

Supplier: Ambeed
Warning

Synonyms: [(R,R)-N-(2-Amino-1,2-diphenylethyl)pentafluorobenzenesulphonamide]chloro(p-cymene)ruthenium(II), Chloro(p-cymene)[(R,R)-N-(pentafluorobenzenesulphonyl)-1,2-diphenylethylenediamine]ruthenium(II)

A472679-250MG A472679-100MG A472679-1G
77613-462EA 44.29 USD
77613-462 77886-229 77886-230
RuCl[(R,R)-Fsdpen](p-cymene) ≥98%
RuCl[(R,R)-Fsdpen](p-cymene)

Precaución: for Research Use Only

Formula: C₃₀H₂₈ClF₅N₂O₂RuS
MW: 712.14 g/mol
MDL Number: MFCD12545963
CAS Number: 1026995-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR