4-Bromo-2-methoxybenzenesulfonyl chloride ≥97%

Supplier: Ambeed
Danger

Synonyms: 4-Bromo-2-methoxybenzene-1-sulphonyl chloride

A472416-1G A472416-250MG A472416-5G
77886-225EA 264 USD
77886-225 77886-226 77886-227
4-Bromo-2-methoxybenzenesulfonyl chloride ≥97%
4-Bromo-2-methoxybenzenesulfonyl chloride

Precaución: for Research Use Only

Formula: C₇H₆BrClO₃S
MDL Number: MFCD11847575
CAS Number: 145915-29-3
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR