1-Bromo-7-methoxynaphthalene ≥95%

Supplier: Ambeed
Warning

A469300-100MG A469300-1G A469300-250MG
77886-217EA 53 USD
77886-217 77886-218 77886-219
1-Bromo-7-methoxynaphthalene ≥95%
1-Bromo-7-methoxynaphthalene

Precaución: for Research Use Only

Formula: C₁₁H₉BrO
MDL Number: MFCD07435364
CAS Number: 83710-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR