Methyl 5-oxopentanoate ≥97%

Supplier: Ambeed
Warning

A469200-250MG A469200-100MG A469200-1G
77613-454EA 60.77 USD
77613-454 77673-200 77673-202
Methyl 5-oxopentanoate ≥97%
Methyl 5-oxopentanoate

Precaución: for Research Use Only

Formula: C₆H₁₀O₃
MW: 130.14 g/mol
MDL Number: MFCD09833062
CAS Number: 6026-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR