5-Bromo-1-methyl-1H-pyrazole-3-carboxylic acid ≥98%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
A469196-1G
A469196-100MG
77684-265EA
392
USD
77684-265
77892-948
5-Bromo-1-methyl-1H-pyrazole-3-carboxylic acid ≥98%, Ambeed.Inc
5-Bromo-1-methyl-1H-pyrazole-3-carboxylic acid
Precaución: For research use only.
Formula:
C₅H₅BrN₂O₂ |
MDL Number:
MFCD22370661 CAS Number: 1222174-93-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...