Dibenzosuberyl chloride ≥98%

Supplier: Ambeed
Danger

Synonyms: 5-Chloro-10,11-dihydro-5H-dibenzo[a,d][7]annulene, 5-Chlorodibenzo[a,d]cycloheptane, 5-Chloro-10,11-dihydro-5H-dibenzo[a,d]cycloheptene

A472139-25G A472139-5G
77602-050EA 246 USD
77602-050 77669-120
Dibenzosuberyl chloride ≥98%
Dibenzosuberyl chloride

Precaución: for Research Use Only

Formula: C₁₅H₁₃Cl
MW: 228.72 g/mol
MDL Number: MFCD00003592
CAS Number: 1210-33-9
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR