3-Phenyl-1H-pyrazol-4-amine ≥95%

Supplier: Ambeed
Warning

Synonyms: 5-Phenyl-1H-pyrazol-4-amine

A468951-250MG A468951-100MG A468951-1G
77684-264EA 83 USD
77684-264 77684-262 77684-263
3-Phenyl-1H-pyrazol-4-amine ≥95%
3-Phenyl-1H-pyrazol-4-amine

Precaución: For research use only.

Formula: C₉H₉N₃
MW: 159.188 g/mol
MDL Number: MFCD01693716
CAS Number: 91857-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR