1-Isopropyl-1H-pyrazole-5-boronic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: (1-Isopropyl-1H-pyrazol-5-yl)boronic acid

A468427-1G A468427-5G
77613-452EA 26.78 USD
77613-452 77684-259
1-Isopropyl-1H-pyrazole-5-boronic acid ≥95%
1-Isopropyl-1H-pyrazole-5-boronic acid

Precaución: For research use only.

Formula: C₆H₁₁BN₂O₂
MW: 153.975 g/mol
MDL Number: MFCD11098002
CAS Number: 839714-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR