(R)-Chroman-3-amine hydrochloride ≥96%
Supplier: Ambeed
|
Warning
|
A471069-100MG
A471069-1G
77886-221EA
81
USD
77886-221
77886-222
(R)-Chroman-3-amine hydrochloride ≥96%
(R)-Chroman-3-amine hydrochloride
Precaución: for Research Use Only
Formula:
C₉H₁₂ClNO MW: 185.65 g/mol |
MDL Number:
MFCD01736331 CAS Number: 211506-59-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...