4-Bromo-1-(oxetan-3-yl)-1H-pyrazole ≥98%

Supplier: Ambeed
Warning

A471011-1G A471011-250MG A471011-5G
77594-484EA 116.85 USD
77594-484 77892-953 77892-954
4-Bromo-1-(oxetan-3-yl)-1H-pyrazole ≥98%
4-Bromo-1-(oxetan-3-yl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O
MDL Number: MFCD21849011
CAS Number: 1374657-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR