(4-Amino-3-fluorophenyl)boronic acid ≥95%

Supplier: Ambeed
Warning

A471005-1G A471005-5G
77890-954EA 90 USD
77890-954 77890-955
(4-Amino-3-fluorophenyl)boronic acid ≥95%
(4-Amino-3-fluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₇BFNO₂
MW: 154.94 g/mol
MDL Number: MFCD10697910
CAS Number: 494752-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR