4-Methylumbelliferyl caprylate ≥98%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-2-oxo-2H-chromen-7-yl octanoate

A467791-250MG A467791-5G
77748-956EA 28 USD
77748-956 77748-957
4-Methylumbelliferyl caprylate ≥98%
4-Methylumbelliferyl caprylate

Precaución: for Research Use Only

Formula: C₁₈H₂₂O₄
MW: 302.365 g/mol
MDL Number: MFCD00038659
CAS Number: 20671-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR