Cefixime ≥97%

Supplier: Ambeed
Danger

Synonyms: (6R,7R)-7-((Z)-2-(2-Aminothiazol-4-yl)-2-((carboxymethoxy)imino)acetamido)-8-oxo-3-vinyl-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid

A467731-50MG A467731-25G A467731-250MG
77594-476EA 5.13 USD
77594-476 77602-042 77602-040
Cefixime ≥97%
Cefixime

Precaución: for Research Use Only

Formula: C₁₆H₁₅N₅O₇S₂
MW: 453.44 g/mol
MDL Number: MFCD00865020
CAS Number: 79350-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR