(5,6-Dimethoxypyridin-3-yl)boronic acid ≥95%

Supplier: Ambeed
Warning

A467007-1G A467007-250MG A467007-5G
77900-631EA 276 USD
77900-631 77900-981 77901-336
(5,6-Dimethoxypyridin-3-yl)boronic acid ≥95%
(5,6-Dimethoxypyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₁₀BNO₄
MDL Number: MFCD18254414
CAS Number: 1346526-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR