(2R,4S,5R,6R)-5-Acetamido-2-(((((2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)-4-hydroxy-6-((1R,2R)-1,2,3-trihydroxypropyl)tetrahydro-2H-pyran-2-carboxylic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: CMP-Sialic acid

A466931-50MG A466931-100MG A466931-1G A466931-250MG
77626-936EA 335.78 USD
77626-936 77878-232 77878-233 77878-234
(2R,4S,5R,6R)-5-Acetamido-2-(((((2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)-4-hydroxy-6-((1R,2R)-1,2,3-trihydroxypropyl)tetrahydro-2H-pyran-2-carboxylic acid ≥98%

Precaución: for Research Use Only

Formula: C₂₀H₃₁N₄O₁₆P
MDL Number: MFCD00870237
CAS Number: 3063-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR