2-Chloro-4-((1-(4-fluorophenyl)-2,5-dimethyl-1H-imidazol-4-yl)ethynyl)pyridine 98+%

Supplier: Ambeed
Warning

Synonyms: Basimglurant

A467450-10MG A467450-1MG A467450-5MG
77613-446EA 35.02 USD
77613-446 77613-448 77613-450
2-Chloro-4-((1-(4-fluorophenyl)-2,5-dimethyl-1H-imidazol-4-yl)ethynyl)pyridine 98+%
2-Chloro-4-((1-(4-fluorophenyl)-2,5-dimethyl-1H-imidazol-4-yl)ethynyl)pyridine

Precaución: for Research Use Only

Formula: C₁₈H₁₃ClFN₃
MW: 325.77 g/mol
MDL Number: MFCD25976719
CAS Number: 802906-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR