(E)-Cyclooct-4-enol 95% rel-(1R-4E-pR)-equatorial

Supplier: Ambeed
Warning

Synonyms: TCO-OH

A467258-250MG A467258-50MG A467258-100MG A467258-1G
77617-022EA 770.44 USD
77617-022 77617-024 77617-020 77748-955
(E)-Cyclooct-4-enol 95% rel-(1R-4E-pR)-equatorial
(E)-Cyclooct-4-enol

Precaución: for Research Use Only

Formula: C₈H₁₄O
MW: 126.19 g/mol
MDL Number: MFCD24386370
CAS Number: 85081-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR