3-(Trifluoromethyl)cyclohexanone ≥97%

Supplier: Ambeed
Warning

A466294-250MG A466294-1G
77617-018EA 49.44 USD
77617-018 77748-952
3-(Trifluoromethyl)cyclohexanone ≥97%
3-(Trifluoromethyl)cyclohexanone

Precaución: for Research Use Only

Formula: C₇H₉F₃O
MW: 166.14 g/mol
MDL Number: MFCD00034662
CAS Number: 585-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR