tert-Butyl 3-(2-(2-(prop-2-yn-1-yloxy)ethoxy)ethoxy)propanoate ≥97%

Supplier: Ambeed
Warning

Synonyms: Propargyl-PEG3-Boc

A458554-250MG A458554-100MG A458554-1G
77616-982EA 112.27 USD
77616-982 77616-978 77616-980
tert-Butyl 3-(2-(2-(prop-2-yn-1-yloxy)ethoxy)ethoxy)propanoate ≥97%
tert-Butyl 3-(2-(2-(prop-2-yn-1-yloxy)ethoxy)ethoxy)propanoate

Precaución: for Research Use Only

Formula: C₁₄H₂₄O₅
MDL Number: MFCD22201522
CAS Number: 1374658-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR