Triethylammonium chloride ≥98%

Supplier: Ambeed
Warning

Synonyms: Triethylamine hydrochloride

A458550-100G A458550-25G A458550-500G
77594-442EA 10.25 USD
77594-442 77594-444 77601-982
Triethylammonium chloride ≥98%
Triethylammonium chloride

Precaución: for Research Use Only

Formula: C₆H₁₆ClN
MDL Number: MFCD00012500
CAS Number: 554-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR