Nec-1s ≥98%

Supplier: Ambeed
Warning

A466111-100MG A466111-1G A466111-250MG
77613-432EA 178.19 USD
77613-432 77892-943 77892-944
Nec-1s ≥98%
Nec-1s

Precaución: for Research Use Only

Formula: C₁₃H₁₂ClN₃O₂
MDL Number: MFCD26743828
CAS Number: 852391-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR