(R)-Piperazin-2-ylmethanol ≥95%

Supplier: Ambeed
Danger

A465962-1G A465962-100MG A465962-250MG
77673-192EA 115 USD
77673-192 77884-192 77884-193
(R)-Piperazin-2-ylmethanol ≥95%
(R)-Piperazin-2-ylmethanol

Precaución: for Research Use Only

Formula: C₅H₁₂N₂O
MW: 116.16 g/mol
MDL Number: MFCD07772089
CAS Number: 149715-47-9
UN: 3259
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR