8-Boc-3-amino-1-oxa-8-azaspiro[4.5]decane ≥98%
Supplier: Ambeed
|
Warning
|
A465689-100MG
A465689-250MG
77882-184EA
70
USD
77882-184
77882-185
8-Boc-3-amino-1-oxa-8-azaspiro[4.5]decane ≥98%
8-Boc-3-amino-1-oxa-8-azaspiro[4.5]decane
Precaución: for Research Use Only
Formula:
C₁₃H₂₄N₂O₃ |
MDL Number:
MFCD12198529 CAS Number: 1160246-91-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...