MorDalPhos Pd G4 ≥98%

Supplier: Ambeed
Warning

A457921-1G A457921-100MG A457921-250MG
77622-312EA 436.72 USD
77622-312 77783-422 77783-423
MorDalPhos Pd G4 ≥98%
MorDalPhos Pd G4

Precaución: for Research Use Only

Formula: C₄₄H₅₇N₂O₄PPdS
MDL Number: MFCD30145864
CAS Number: 2230788-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR