Methyl 5-iodo-1-methyl-1H-pyrazole-3-carboxylate ≥95%

Supplier: Ambeed
Warning

A457654-1G A457654-250MG
77673-160EA 498 USD
77673-160 77673-162
Methyl 5-iodo-1-methyl-1H-pyrazole-3-carboxylate ≥95%
Methyl 5-iodo-1-methyl-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇IN₂O₂
MDL Number: MFCD16660450
CAS Number: 1193244-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR