2,4-Dicyanoaniline ≥98%

Supplier: Ambeed
Danger

A465204-1G A465204-250MG
77890-946EA 45 USD
77890-946 77890-947
2,4-Dicyanoaniline ≥98%
2,4-Dicyanoaniline

Precaución: for Research Use Only

Formula: C₈H₅N₃
MDL Number: MFCD00270798
CAS Number: 19619-22-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR