N-Acetyl-D-methionine ≥99%

Supplier: Ambeed
Warning

Synonyms: Ac-D-Met-OH, N-Ac-D-Met-OH, (2R)-2-acetamido-4-(methylsulfanyl)butanoic acid, N-Acetyl-D-Methionine

A465068-25G A465068-5G
77626-930EA 122.57 USD
77626-930 77626-932
N-Acetyl-D-methionine ≥99%
N-Acetyl-D-methionine

Precaución: for Research Use Only

Formula: C₇H₁₃NO₃S
MW: 191.2 g/mol
MDL Number: MFCD00069962
CAS Number: 1509-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR