3-(2-Methoxyethoxy)benzoic acid ≥95%

Supplier: Ambeed
Warning

A457382-100MG A457382-1G
77890-930EA 51 USD
77890-930 77890-931
3-(2-Methoxyethoxy)benzoic acid ≥95%
3-(2-Methoxyethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄
MDL Number: MFCD09036534
CAS Number: 152808-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR